missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Phenylbenzimidazole-5-sulfonic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 259700050
Specifications
2-Phenylbenzimidazole-5-sulfonic acid | |
Glass bottle | |
White | |
5g | |
97.5 | |
98% | |
MFCD00053007 | |
1h-benzimidazole-5-sulfonic acid, 2-phenyl, 2-phenyl-1h-benzimidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 2-phenyl-5-benzimidazolesulfonic acid, 2-phenylbenzimidazole-5-sulfonic acid, ensulizole, parsol hs, phenylbenzimidazole sulfonic acid, unii-9yq9di1w42 | |
UVCJGUGAGLDPAA-UHFFFAOYSA-N | |
2-phenyl-3H-benzimidazole-5-sulfonic acid | |
33919 | |
Fine Crystalline Powder |
Authentic | |
7% max. | |
>300°C | |
27503-81-7 | |
100.0 | |
C13H10N2O3S | |
15, 3647 | |
Solubility in water: soluble | |
C1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)S(=O)(=O)O | |
274.28 | |
274.28 | |
98% |
Chemical Identifiers
27503-81-7 | |
274.28 | |
UVCJGUGAGLDPAA-UHFFFAOYSA-N | |
33919 | |
C1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)S(=O)(=O)O |
C13H10N2O3S | |
MFCD00053007 | |
1h-benzimidazole-5-sulfonic acid, 2-phenyl, 2-phenyl-1h-benzimidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 2-phenyl-5-benzimidazolesulfonic acid, 2-phenylbenzimidazole-5-sulfonic acid, ensulizole, parsol hs, phenylbenzimidazole sulfonic acid, unii-9yq9di1w42 | |
2-phenyl-3H-benzimidazole-5-sulfonic acid |
Safety and Handling
EINECSNumber : 248-502-