Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenylanthraquinone, 98%

$50.34 - $118.73

Chemical Identifiers

Chemical Name or Material 2-Phenylanthraquinone
CAS 6485-97-8
Molecular Formula C20H12O2
Formula Weight 284.32
MDL Number MFCD00559262
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2737603

  SDS
alfa aesar™
H2737603
1g Each for $118.73

AAH27376MD

  SDS
alfa aesar™
H27376MD
250mg Each for $50.34
Description & Specifications

Specifications

Formula Weight 284.32
MDL Number MFCD00559262
Melting Point 160° to 163°C
Quantity 1g
Molecular Formula C20H12O2
Assay 98%
CAS 6485-97-8
Chemical Name or Material 2-Phenylanthraquinone