missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenyl-L-glycinamide hydrochloride, 97%

$68.31 - $309.96

Chemical Identifiers

CAS 60079-51-8
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.639
MDL Number MFCD00270568
InChI Key LXOAFHGMXCFTOU-FJXQXJEOSA-N
Synonym s-2-amino-2-phenylacetamide hydrochloride, l-phenylglycine amide hydrochloride, h-phg-nh2 hcl, h-phg-nh2.hcl, 2s-2-amino-2-phenylacetamide hydrochloride, hpphg-nh2 hcl, h-phg-nh2 . hcl, phenylglycine amide hydrochloride, s-2-amino-2-phenyl-acetamide hydrochloride, alphas-alpha-amino-benzeneacetamide hydrochloride
PubChem CID 22850726
IUPAC Name (2S)-2-amino-2-phenylacetamide;hydrochloride
SMILES C1=CC=C(C=C1)C(C(=O)N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6575103 View Documents Alfa Aesar™
H6575103
1g Each for $68.31
 
AAH6575106 View Documents Alfa Aesar™
H6575106
5g Each for $309.96
 
Specifications

Chemical Identifiers

60079-51-8
186.639
LXOAFHGMXCFTOU-FJXQXJEOSA-N
22850726
C1=CC=C(C=C1)C(C(=O)N)N.Cl
C8H11ClN2O
MFCD00270568
s-2-amino-2-phenylacetamide hydrochloride, l-phenylglycine amide hydrochloride, h-phg-nh2 hcl, h-phg-nh2.hcl, 2s-2-amino-2-phenylacetamide hydrochloride, hpphg-nh2 hcl, h-phg-nh2 . hcl, phenylglycine amide hydrochloride, s-2-amino-2-phenyl-acetamide hydrochloride, alphas-alpha-amino-benzeneacetamide hydrochloride
(2S)-2-amino-2-phenylacetamide;hydrochloride

Specifications

1g
C8H11ClN2O
4823667
LXOAFHGMXCFTOU-FJXQXJEOSA-N
(2S)-2-amino-2-phenylacetamide;hydrochloride
22850726
Solid
2-Phenyl-L-glycinamide hydrochloride
60079-51-8
MFCD00270568
s-2-amino-2-phenylacetamide hydrochloride, l-phenylglycine amide hydrochloride, h-phg-nh2 hcl, h-phg-nh2.hcl, 2s-2-amino-2-phenylacetamide hydrochloride, hpphg-nh2 hcl, h-phg-nh2 . hcl, phenylglycine amide hydrochloride, s-2-amino-2-phenyl-acetamide hydrochloride, alphas-alpha-amino-benzeneacetamide hydrochloride
C1=CC=C(C=C1)C(C(=O)N)N.Cl
186.639
186.64
97%