Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenyl-4-penten-2-ol, 98%

$45.81 - $45.81

Chemical Identifiers

Chemical Name or Material 2-Phenyl-4-penten-2-ol
CAS 4743-74-2
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00060886
Synonym Allyl methyl phenyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2024806

  SDS
alfa aesar™
B2024806
5g Each for $45.81
Description & Specifications

Specifications

Formula Weight 162.23
Synonym Allyl methyl phenyl carbinol
Assay 98%
Molecular Formula C11H14O
CAS 4743-74-2
Quantity 5g
MDL Number MFCD00060886
Chemical Name or Material 2-Phenyl-4-penten-2-ol