Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenyl-2-pentanol, 98%

$39.11 - $554.14

Chemical Identifiers

Chemical Name or Material 2-Phenyl-2-pentanol
CAS 4383-18-0
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00046592
Synonym Methyl phenyl n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2085403

  SDS
alfa aesar™
B2085403
1g Each for $39.11

AAB2085406

  SDS
alfa aesar™
B2085406
5g Each for $138.54

AAB2085414

  SDS
alfa aesar™
B2085414
25g Each for $554.14
Description & Specifications

Specifications

Formula Weight 164.25
Chemical Name or Material 2-Phenyl-2-pentanol
Assay 98%
Molecular Formula C11H16O
MDL Number MFCD00046592
CAS 4383-18-0
Synonym Methyl phenyl n-propyl carbinol
Quantity 1g