Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenyl-1-propylboronic acid pinacol ester, 97%

$109.85 - $988.43

Chemical Identifiers

Chemical Name or Material 2-Phenyl-1-propylboronic acid pinacol ester
CAS 280559-30-0
Molecular Formula C15H23BO2
Formula Weight 246.16
MDL Number MFCD09953475
Synonym 4,4,5,5-Tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2693103

  SDS
alfa aesar™
H2693103
1g Each for $323.96

AAH2693106

  SDS
alfa aesar™
H2693106
5g Each for $988.43

AAH26931MD

  SDS
alfa aesar™
H26931MD
250mg Each for $109.85
Description & Specifications

Specifications

Formula Weight 246.16
MDL Number MFCD09953475
Chemical Name or Material 2-Phenyl-1-propylboronic acid pinacol ester
Molecular Formula C15H23BO2
CAS 280559-30-0
Quantity 1g
Assay 97%
Synonym 4,4,5,5-Tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane