Please login to your online account to display your discounted pricing

2-Phenyl-1,3-thiazole-4-carbonyl chloride, Maybridge

$109.89 - $109.89

Chemical Identifiers

Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carbonyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H6ClNOS
Formula Weight 223.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC18302DA

  SDS
fisher scientific company
CC18302DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC18302DE

 
fisher scientific company
CC18302DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC18302CB

  SDS
fisher scientific company
CC18302CB
Each for $109.89
Description & Specifications

Specifications

Formula Weight 223.68
Quantity 1g
Assay 95%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carbonyl chloride
CAS 36094-04-9
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C10H6ClNOS