missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Phenyl-1,3-thiazole-4-carbonyl chloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC18302DA
Specifications
2-Phenyl-1, 3-thiazole-4-carbonyl chloride | |
95% | |
C10H6ClNOS | |
2-phenylthiazole-4-carbonyl chloride, 2-phenyl-thiazole-4-carbonyl chloride, 4-thiazolecarbonylchloride, 2-phenyl | |
C1=CC=C(C=C1)C2=NC(=CS2)C(=O)Cl | |
223.674 | |
223.68 |
36094-04-9 | |
Amber Glass Bottle | |
1g | |
ZZFOIDMEHXGBKS-UHFFFAOYSA-N | |
2-phenyl-1,3-thiazole-4-carbonyl chloride | |
2776472 | |
95% |