missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Phenyl-1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

$179.36 - $179.36

Chemical Identifiers

CAS 165736-03-8
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD02682055
InChI Key PMFCMLGAPHZRTE-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
PubChem CID 736526
IUPAC Name (2-phenyl-1,3-thiazol-4-yl)methanamine
SMILES C1=CC=C(C=C1)C2=NC(=CS2)CN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC18313DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC18313DA
1 g Amber Glass Bottle
Each for $179.36
 
Description

Description

Specifications

Chemical Identifiers

165736-03-8
190.264
PMFCMLGAPHZRTE-UHFFFAOYSA-N
736526
C1=CC=C(C=C1)C2=NC(=CS2)CN
C10H10N2S
MFCD02682055
2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
(2-phenyl-1,3-thiazol-4-yl)methanamine

Specifications

165736-03-8
100.0
Amber Glass Bottle
MFCD02682055
2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
C1=CC=C(C=C1)C2=NC(=CS2)CN
190.264
190.27
(2-Phenyl-1, 3-thiazol-4-yl)methylamine
96.5
97%
C10H10N2S
1 g
PMFCMLGAPHZRTE-UHFFFAOYSA-N
(2-phenyl-1,3-thiazol-4-yl)methanamine
736526
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'