missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Phenyl-1,3-thiazol-4-yl)methylamine, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC18313EA

 View more versions of this product

Catalog No. CC18313EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(2-Phenyl-1, 3-thiazol-4-yl)methylamine
10g
96.5
97%
MFCD02682055
PMFCMLGAPHZRTE-UHFFFAOYSA-N
(2-phenyl-1,3-thiazol-4-yl)methanamine
736526
97%
Amber Glass Bottle
165736-03-8
100.0
C10H10N2S
1-2-phenyl-1,3-thiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, 2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 4-aminomethyl-2-phenyl-1,3-thiazole, 4-thiazolemethanamine, 2-phenyl, c-2-phenyl-thiazol-4-yl-methylamine, chembrdg-bb 4101065, enamine_005991
C1=CC=C(C=C1)C2=NC(=CS2)CN
190.264
190.27

Chemical Identifiers

165736-03-8
190.264
PMFCMLGAPHZRTE-UHFFFAOYSA-N
736526
C1=CC=C(C=C1)C2=NC(=CS2)CN
C10H10N2S
MFCD02682055
1-2-phenyl-1,3-thiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, 2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 4-aminomethyl-2-phenyl-1,3-thiazole, 4-thiazolemethanamine, 2-phenyl, c-2-phenyl-thiazol-4-yl-methylamine, chembrdg-bb 4101065, enamine_005991
(2-phenyl-1,3-thiazol-4-yl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents