missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxythioacetamide, 97%

$195.49 - $195.49

Chemical Identifiers

CAS 35370-80-0
Molecular Formula C8H9NOS
Formula Weight 167.26
MDL Number MFCD00100035
Synonym 2-Phenoxyethanethioamide
Chemical Name or Material 2-Phenoxythioacetamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2647803 SDS Alfa Aesar™
H2647803
1g Each for $195.49
AAH26478MD SDS Alfa Aesar™
H26478MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

35370-80-0
167.26
2-Phenoxyethanethioamide
C8H9NOS
MFCD00100035
2-Phenoxythioacetamide

Specifications

114° to 116°C
35370-80-0
167.26
2-Phenoxyethanethioamide
97%
1g
C8H9NOS
MFCD00100035
2-Phenoxythioacetamide

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger