missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxypropylamine, 98%

CAS: 6437-49-6 | C9H14ClNO | 187.667 g/mol

Supplier:  Thermo Scientific Chemicals H5072003

 View more versions of this product

Catalog No. AAH5072003

Only null left
Explore available promotions

Description

Description

2-Phenoxypropylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxypropylamine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Specifications

Specifications

2-Phenoxypropylamine
C9H14ClNO
1 g
Air Sensitive
Insoluble in water.
CC(CN)OC1=CC=CC=C1.Cl
187.667
151.21
6437-49-6
MFCD07781046
UN3259
2-phenoxypropylamine hydrochloride, 2-phenoxypropan-1-amine hydrochloride, 2-phenoxypropylamine hcl, 1-aminopropan-2-yl oxy benzene hydrochloride, acmc-20annz, 2-phenoxypropan-1-amine-hydrogen chloride 1/1
CGOBBGUREQJPPH-UHFFFAOYSA-N
2-phenoxypropan-1-amine;hydrochloride
24212021
98%

Chemical Identifiers

6437-49-6
187.667
CGOBBGUREQJPPH-UHFFFAOYSA-N
24212021
CC(CN)OC1=CC=CC=C1.Cl
C9H14ClNO
MFCD07781046
2-phenoxypropylamine hydrochloride, 2-phenoxypropan-1-amine hydrochloride, 2-phenoxypropylamine hcl, 1-aminopropan-2-yl oxy benzene hydrochloride, acmc-20annz, 2-phenoxypropan-1-amine-hydrogen chloride 1/1
2-phenoxypropan-1-amine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only