Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxyphenylacetonitrile, 98%

$96.51 - $384.82

Chemical Identifiers

Chemical Name or Material 2-Phenoxyphenylacetonitrile
CAS 25562-98-5
Molecular Formula C14H11NO
Formula Weight 209.25
MDL Number MFCD00016386
Beilstein 2727468
Synonym 2-Phenoxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1019403

 
alfa aesar™
L1019403
1g Each for $96.51

AAL1019406

 
alfa aesar™
L1019406
5g Each for $384.82
Description & Specifications

Specifications

Formula Weight 209.25
Assay 98%
Chemical Name or Material 2-Phenoxyphenylacetonitrile
Molecular Formula C14H11NO
MDL Number MFCD00016386
CAS 25562-98-5
Beilstein 2727468
Synonym 2-Phenoxybenzyl cyanide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Quantity 1g
Flash Point >110°C (230°F)
Boiling Point 145°C (1mmHg)
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.