Please login to your online account to display your discounted pricing

2-Phenoxynicotinic acid, 98%, Alfa Aesar™

$98.66 - $98.66

Chemical Identifiers

Chemical Name or Material 2-Phenoxynicotinic acid
CAS 35620-71-4
Molecular Formula C12H9NO3
Formula Weight 215.21
MDL Number MFCD00014629
Beilstein 172906
Synonym 2-Phenoxypyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1124706

 
alfa aesar
A1124706
Each for $98.66
Description & Specifications

Specifications

Formula Weight 215.21
Melting Point 179° to 182°C
Quantity 5g
Chemical Name or Material 2-Phenoxynicotinic acid
Assay 98%
Molecular Formula C12H9NO3
MDL Number MFCD00014629
CAS 35620-71-4
Beilstein 172906
Synonym 2-Phenoxypyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.