missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Phenoxymethyl)benzylamine, 97%, Maybridge

Chemical Identifiers

CAS 168971-56-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C14H15NO
Formula Weight 213.28
MDL Number MFCD04108110
Chemical Name or Material 2-(Phenoxymethyl)benzylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC58513EA Fisher Scientific Company
CC58513EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58513DA SDS Fisher Scientific Company
CC58513DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

168971-56-0
100.0
C14H15NO
MFCD04108110
97%
96.5
97%
213.28
2-(Phenoxymethyl)benzylamine

Specifications

10g
96.5
97%
213.28
2-(Phenoxymethyl)benzylamine
168971-56-0
100.0
C14H15NO
MFCD04108110
97%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.