Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Phenoxymethyl)benzoic acid, 98%

$41.11 - $603.39

Chemical Identifiers

Chemical Name or Material 2-(Phenoxymethyl)benzoic acid
CAS 724-98-1
Molecular Formula C14H12O3
Formula Weight 228.25
MDL Number MFCD00020294
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2519403

  SDS
alfa aesar™
B2519403
1g Each for $41.11

AAB2519406

  SDS
alfa aesar™
B2519406
5g Each for $148.85

AAB2519414

  SDS
alfa aesar™
B2519414
25g Each for $603.39
Description & Specifications

Specifications

Formula Weight 228.25
Chemical Name or Material 2-(Phenoxymethyl)benzoic acid
TSCA TSCA
Assay 98%
Molecular Formula C14H12O3
MDL Number MFCD00020294
CAS 724-98-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.