Please login to your online account to display your discounted pricing

2-Phenoxyethylamine, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Phenoxyethylamine
CAS 1758-46-9
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H11NO
Linear Formula C6H5O(CH2)2NH2
Formula Weight 137.18
Synonym 1-Amino-2-phenoxyethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03254EA

  SDS
fisher scientific company
RF03254EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.18
Quantity 10g
Assay 95%
Density 1.0480g/mL
CAS Min % 95.0
Linear Formula C6H5O(CH2)2NH2
Synonym 1-Amino-2-phenoxyethane
Chemical Name or Material 2-Phenoxyethylamine
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C8H11NO
Packaging Amber glass bottle
Health Hazard 1 Danger
Assay Percent Range 95%
CAS 1758-46-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.