Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxyethyl chloride, 97%

$47.41 - $47.41

Chemical Identifiers

Chemical Name or Material 2-Phenoxyethyl chloride
CAS 622-86-6
Molecular Formula C6H5O(CH2)2Cl
Formula Weight 156.61
MDL Number MFCD00018946
Beilstein 971048
Synonym 2-Chloroethyl phenyl ether; beta-Chlorophenetole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1134506

 
alfa aesar™
L1134506
5g Each for $47.41
Description & Specifications

Specifications

Formula Weight 156.61
Chemical Name or Material 2-Phenoxyethyl chloride
Assay 97%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA TSCA
MDL Number MFCD00018946
CAS 622-86-6
Beilstein 971048
Synonym 2-Chloroethyl phenyl ether; beta-Chlorophenetole
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Flash Point 109°C (228°F)
Boiling Point 88° to 93°C (15mmHg)
Melting Point 23° to 25°C
Molecular Formula C6H5O(CH2)2Cl
Health Hazard 1 Warning
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.