missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxyethyl chloride, 97%

Manufacturer:  Alfa Aesar™ L1134514

 View more versions of this product

Catalog No. AAL1134514



Specifications

Specifications

2-Phenoxyethyl chloride
88°C to 93°C (15mmHg)
25g
C8H9ClO
971048
VQUYNUJARXBNPK-UHFFFAOYSA-N
2-chloroethoxybenzene
12156
97%
109°C (228°F)
23°C to 25°C
622-86-6
MFCD00018946
2-chloroethoxy benzene, beta-chlorophenetole, 2-phenoxyethyl chloride, benzene, 2-chloroethoxy, 2-chloroethyl phenyl ether, b-chlorophenetole, phenetole, .beta.-chloro, phenoxyethyl chloride, benzene, chloroethoxy, 2-chloroethyoxy benzene
C1=CC=C(C=C1)OCCCl
156.609
156.61

Chemical Identifiers

622-86-6
156.609
VQUYNUJARXBNPK-UHFFFAOYSA-N
12156
C1=CC=C(C=C1)OCCCl
C8H9ClO
MFCD00018946
2-chloroethoxy benzene, beta-chlorophenetole, 2-phenoxyethyl chloride, benzene, 2-chloroethoxy, 2-chloroethyl phenyl ether, b-chlorophenetole, phenetole, .beta.-chloro, phenoxyethyl chloride, benzene, chloroethoxy, 2-chloroethyoxy benzene
2-chloroethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

Documents