Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxyethyl acrylate, tech. 85%, stab. with 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material 2-Phenoxyethyl acrylate
CAS 48145-04-6
Name Note stabilized with 4-methoxyphenol
Purity Grade Technical Grade
Molecular Formula C11H12O3
Linear Formula C6H5O(CH2)2O2CCH=CH2
Formula Weight 192.22
MDL Number MFCD00053696
UN Number UN3082
Beilstein 2102773
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1212018

  SDS
alfa aesar™
L1212018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.22
MDL Number MFCD00053696
Linear Formula C6H5O(CH2)2O2CCH=CH2
UN Number UN3082
Boiling Point 103° to 104°C (1mmHg)
Recommended Storage 2° to 8°C
Chemical Name or Material 2-Phenoxyethyl acrylate
CAS 48145-04-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym Acrylic acid 2-phenoxyethyl ester; Ethylene glycol phenyl ether acrylate
Health Hazard 1 Warning
Assay 85%
Purity Grade Technical Grade
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Beilstein 2102773
TSCA TSCA
Name Note stabilized with 4-methoxyphenol
RTECS Number KM0700000
Flash Point >110°C (230°F)
Quantity 50g
Molecular Formula C11H12O3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C