missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxyethyl acrylate, tech. 85%, stab. with 4-methoxyphenol

Manufacturer:  Alfa Aesar™ L1212018

Catalog No. AAL1212018



Description

Description

Specifications

Specifications

2-Phenoxyethyl acrylate
>110°C (230°F)
103°C to 104°C (1mmHg)
stabilized with 4-methoxyphenol
C11H12O3
MFCD00053696
2102773
RZVINYQDSSQUKO-UHFFFAOYSA-N
2-phenoxyethyl prop-2-enoate
39485
85%
1.104
1.519
50g
48145-04-6
C6H5O(CH2)2O2CCH=CH2
UN3082
2-phenoxyethanol acrylate, 2-phenoxyethyl acrylate, 2-propenoic acid, 2-phenoxyethyl ester, acrylic acid, 2-phenoxyethyl ester, chemlink 160, ebecryl 110, ethylene glycol phenyl ether acrylate, light ester po-a, phenoxyethyl acrylate, phenyl cellosolve acrylate
C=CC(=O)OCCOC1=CC=CC=C1
192.214
192.22
Technical

Chemical Identifiers

48145-04-6
192.214
RZVINYQDSSQUKO-UHFFFAOYSA-N
39485
C=CC(=O)OCCOC1=CC=CC=C1
C11H12O3
MFCD00053696
2-phenoxyethanol acrylate, 2-phenoxyethyl acrylate, 2-propenoic acid, 2-phenoxyethyl ester, acrylic acid, 2-phenoxyethyl ester, chemlink 160, ebecryl 110, ethylene glycol phenyl ether acrylate, light ester po-a, phenoxyethyl acrylate, phenyl cellosolve acrylate
2-phenoxyethyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H361fd

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 256-360-6

RTECSNumber : KM0700000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents