Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxybiphenyl, 98+%

$52.59 - $191.36

Chemical Identifiers

Chemical Name or Material 2-Phenoxybiphenyl
CAS 6738-04-1
Molecular Formula C18H14O
Formula Weight 246.31
MDL Number MFCD00003035
Beilstein 2371408
Synonym 2-Biphenylyl phenyl ether; 2-Phenyldiphenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0618306

  SDS
alfa aesar™
L0618306
5g Each for $52.59

AAL0618314

  SDS
alfa aesar™
L0618314
25g Each for $191.36
Description & Specifications

Specifications

Formula Weight 246.31
Chemical Name or Material 2-Phenoxybiphenyl
Assay ≥98%
Molecular Formula C18H14O
MDL Number MFCD00003035
CAS 6738-04-1
Beilstein 2371408
Synonym 2-Biphenylyl phenyl ether; 2-Phenyldiphenyl ether
Quantity 5g
Melting Point 47° to 50°C