missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxybenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5326403

 View more versions of this product

Catalog No. AAH5326403



Description

Description

Applications
Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Phenoxybenzeneboronic acid
1g
C12H11BO3
2-phenoxybenzeneboronic acid, 2-phenoxyphenyl boronic acid, 2-phenoxyphenyl boranediol, 2-phenoxy phenylboronic acid, boronic acid, 2-phenoxyphenyl, pubchem9521, acmc-1bnrk, ksc174e3r, 2-phenoxy-phenyl boronic acid, 2-phenoxyphenylboronicacid
B(C1=CC=CC=C1OC2=CC=CC=C2)(O)O
214.027
214.03
116°C
108238-09-1
MFCD01001592
AVOWPOFIQZSVGV-UHFFFAOYSA-N
(2-phenoxyphenyl)boronic acid
2773559
98%

Chemical Identifiers

108238-09-1
214.027
AVOWPOFIQZSVGV-UHFFFAOYSA-N
2773559
B(C1=CC=CC=C1OC2=CC=CC=C2)(O)O
C12H11BO3
MFCD01001592
2-phenoxybenzeneboronic acid, 2-phenoxyphenyl boronic acid, 2-phenoxyphenyl boranediol, 2-phenoxy phenylboronic acid, boronic acid, 2-phenoxyphenyl, pubchem9521, acmc-1bnrk, ksc174e3r, 2-phenoxy-phenyl boronic acid, 2-phenoxyphenylboronicacid
(2-phenoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents