Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Phenoxyacetamide, 98%

$48.97 - $48.97

Chemical Identifiers

Chemical Name or Material 2-Phenoxyacetamide
CAS 621-88-5
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00017140
Beilstein 1941781
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1493406

  SDS
alfa aesar™
A1493406
5g Each for $48.97

AAA1493414

  SDS
alfa aesar™
A1493414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Phenoxyacetamide
Quantity 5g
CAS 621-88-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 98%
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00017140
Beilstein 1941781
Melting Point 100° to 104°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.