missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Pentynoic acid, 97%

Manufacturer:  Alfa Aesar™ H5341114

 View more versions of this product

Catalog No. AAH5341114



Description

Description

Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Pentynoic acid
47°C to 53°C
5963-77-9
CH3CH2CH≡CCO2H
2-pentynoic acid, pentynoic acid, 2-pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s, 2-pentynoic acid
CCC#CC(=O)O
98.101
98.1
96°C (205°F)
25g
C5H6O2
MFCD00014370
MINRDQDGBLQBGD-UHFFFAOYSA-N
pent-2-ynoic acid
324379
97%

Chemical Identifiers

5963-77-9
98.101
MINRDQDGBLQBGD-UHFFFAOYSA-N
324379
CCC#CC(=O)O
C5H6O2
MFCD00014370
2-pentynoic acid, pentynoic acid, 2-pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s, 2-pentynoic acid
pent-2-ynoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents