missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Pentanone, 99%, pure, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC123500100
Specifications
2-Pentanone | |
7°C | |
Plastic drum | |
0.809 | |
0.1% Max. (K.F.) | |
-78.0°C | |
107-87-9 | |
C5H10O | |
01,676 | |
2-pentanone, ethyl acetone, ethylacetone, methyl n-propyl ketone, methyl propyl ketone, methyl-propyl-cetone, methylpropyl ketone, metylopropyloketon, pentanone, propyl methyl ketone | |
XNLICIUVMPYHGG-UHFFFAOYSA-N | |
pentan-2-one | |
7895 | |
86.13 | |
99% |
0.8090g/mL | |
Authentic | |
1.3890 to 1.3910 | |
0.5 mPa.s (20°C) | |
100.0°C to 103.0°C | |
10L | |
98.5% min. (GC) | |
CH3CO(CH2)CH3 | |
15,6187 | |
Solubility in water: 43g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCCC(=O)C | |
86.13 | |
CHEBI:16472 | |
Liquid | |
Pure |
Chemical Identifiers
107-87-9 | |
86.13 | |
2-pentanone, ethyl acetone, ethylacetone, methyl n-propyl ketone, methyl propyl ketone, methyl-propyl-cetone, methylpropyl ketone, metylopropyloketon, pentanone, propyl methyl ketone | |
CHEBI:16472 | |
CCCC(=O)C |
C5H10O | |
XNLICIUVMPYHGG-UHFFFAOYSA-N | |
7895 | |
pentan-2-one |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 203-528-1