Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(p-Tolyl)pyridine, 98%

$69.00 - $195.88

Chemical Identifiers

Chemical Name or Material 2-(p-Tolyl)pyridine
CAS 4467-06-5
Molecular Formula C12H11N
Formula Weight 169.22
MDL Number MFCD00006281
Synonym 2-(4-Methylphenyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2657406

  SDS
alfa aesar™
H2657406
5g Each for $69.00

AAH2657414

  SDS
alfa aesar™
H2657414
25g Each for $195.88
Description & Specifications

Specifications

Formula Weight 169.22
CAS 4467-06-5
Flash Point >110°C (230°F)
Boiling Point 170° to 180°C (20mmHg)
Chemical Name or Material 2-(p-Tolyl)pyridine
Molecular Formula C12H11N
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
MDL Number MFCD00006281
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Synonym 2-(4-Methylphenyl)pyridine
Assay 98%
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.