missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(p-Tolyl)benzoic Acid, TCI America™

Manufacturer:  TCI America T237725G

 View more versions of this product

Catalog No. T237725G



Specifications

Specifications

2-(p-Tolyl)benzoic Acid
150°C
7148-03-0
MFCD00045826
ZSTUEICKYWFYIC-UHFFFAOYSA-N
2-(4-methylphenyl)benzoic acid
81568
Crystal-Powder at 20°C
White-Yellow
25g
C14H12O2
2-(4-Methylphenyl)benzoic Acid, 4′-Methylbiphenyl-2-carboxylic Acid
CC1=CC=C(C=C1)C2=CC=CC=C2C(=O)O
212.248
212.25
≥98.0% (GC,T)

Chemical Identifiers

7148-03-0
212.248
ZSTUEICKYWFYIC-UHFFFAOYSA-N
81568
CC1=CC=C(C=C1)C2=CC=CC=C2C(=O)O
C14H12O2
MFCD00045826
2-(4-Methylphenyl)benzoic Acid, 4′-Methylbiphenyl-2-carboxylic Acid
2-(4-methylphenyl)benzoic acid
Safety and Handling

Safety and Handling

TSCA : Yes

Documents