Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester, 97%

$65.09 - $213.96

Chemical Identifiers

Chemical Name or Material 2-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester
CAS 796061-07-9
Molecular Formula C19H24BNO4S
Formula Weight 373.27
MDL Number MFCD05663862
Synonym 4-Methyl-N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2725003

  SDS
alfa aesar™
H2725003
1g Each for $65.09

AAH2725006

  SDS
alfa aesar™
H2725006
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 373.27
Molecular Formula C19H24BNO4S
Chemical Name or Material 2-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester
Melting Point 135° to 139°C
Quantity 1g
Synonym 4-Methyl-N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide
MDL Number MFCD05663862
CAS 796061-07-9
Assay 97%