Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(p-Toluenesulfonyl)ethanol, 98%

$53.73 - $53.73

Chemical Identifiers

Chemical Name or Material 2-(p-Toluenesulfonyl)ethanol
CAS 22381-54-0
Molecular Formula C9H12O3S
Formula Weight 200.26
MDL Number MFCD00041757
Beilstein 1961132
Synonym 2-(4-Methylphenylsulfonyl)ethanol; 2-(p-Tosyl)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1824903

 
alfa aesar™
A1824903
1g Each for $53.73
Description & Specifications

Specifications

Formula Weight 200.26
TSCA TSCA
Chemical Name or Material 2-(p-Toluenesulfonyl)ethanol
Assay 98%
Molecular Formula C9H12O3S
MDL Number MFCD00041757
CAS 22381-54-0
Beilstein 1961132
Synonym 2-(4-Methylphenylsulfonyl)ethanol; 2-(p-Tosyl)ethanol
Boiling Point 178° to 181°C (0.2mmHg)
Melting Point 50° to 53°C
Quantity 1g

Applications
2-(p-Toluenesulfonyl)ethanol is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.