missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(p-Toluenesulfonyl)acetic Acid 98.0+%, TCI America™
Supplier: TCI America T241625G
Specifications
2-(p-Toluenesulfonyl)acetic Acid | |
118°C | |
MFCD00021764 | |
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o | |
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O | |
214.235 | |
214.24 | |
Crystalline Powder |
3937-96-0 | |
C9H10O4S | |
25 g | |
AQDHXMBUTDLAMD-UHFFFAOYSA-N | |
2-(4-methylphenyl)sulfonylacetic acid | |
77538 | |
≥98.0% (HPLC) |
Chemical Identifiers
3937-96-0 | |
214.235 | |
AQDHXMBUTDLAMD-UHFFFAOYSA-N | |
77538 | |
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O |
C9H10O4S | |
MFCD00021764 | |
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o | |
2-(4-methylphenyl)sulfonylacetic acid |
Safety and Handling
TSCA : No