Learn More
2-Oxoindoline-5-sulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC53303DA
Specifications
2-Oxoindoline-5-sulfonyl chloride | |
80.0 | |
97% | |
C8H6ClNO3S | |
1 g | |
FYBNVKMJOOYPGI-UHFFFAOYSA-N | |
2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride | |
4962506 | |
Tech. |
199328-31-9 | |
100.0 | |
Amber Glass Bottle | |
MFCD06655924 | |
2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole | |
ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1 | |
231.65 | |
231.66 | |
Technical |
Chemical Identifiers
199328-31-9 | |
231.65 | |
FYBNVKMJOOYPGI-UHFFFAOYSA-N | |
4962506 |
C8H6ClNO3S | |
MFCD06655924 | |
2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole | |
ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1 |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger