Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Oxocyclopentylacetic acid, 97%

$64.65 - $197.03

Chemical Identifiers

Chemical Name or Material 2-Oxocyclopentylacetic acid
CAS 1460-38-4
Molecular Formula C7H10O3
Formula Weight 142.15
MDL Number MFCD02093672
Synonym Cyclopentanone-2-acetic acid; 2-Oxocyclopentaneacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2697703

  SDS
alfa aesar™
H2697703
1g Each for $64.65

AAH2697706

  SDS
alfa aesar™
H2697706
5g Each for $197.03
Description & Specifications

Specifications

Formula Weight 142.15
Chemical Name or Material 2-Oxocyclopentylacetic acid
Assay 97%
Molecular Formula C7H10O3
MDL Number MFCD02093672
CAS 1460-38-4
Synonym Cyclopentanone-2-acetic acid; 2-Oxocyclopentaneacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 47° to 51°C
Flash Point >110°C (230°F)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.