Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Oxocyclohexanepropionitrile, 99%

Chemical Identifiers

Chemical Name or Material 2-Oxocyclohexanepropionitrile
CAS 4594-78-9
Molecular Formula C9H13NO
Formula Weight 151.21
MDL Number MFCD00013797
UN Number UN3276
Synonym 2-(beta-Cyanoethyl)cyclohexanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5072203

  SDS
alfa aesar™
H5072203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.21
UN Number UN3276
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 2-Oxocyclohexanepropionitrile
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 2-(beta-Cyanoethyl)cyclohexanone
Health Hazard 1 Danger
Molecular Formula C9H13NO
Assay 99%
Quantity 1g
Boiling Point 138° to 142°C (10mmHg)
MDL Number MFCD00013797
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
CAS 4594-78-9
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.