Learn More
2-Oxocyclohexanepropionitrile, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5072206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Oxocyclohexanepropionitrile | |
1.021 | |
C9H13NO | |
5 g | |
3-2-oxocyclohexyl propanenitrile, 2-2-cyanoethyl cyclohexanone, cyclohexanepropanenitrile, 2-oxo, 2-2-cyanethyl cyclohexanone, 2-oxo-1-cyclohexanepropionitrile, cyclohexanepropionitrile, 2-oxo, 2-oxocyclohexanepropionitrile, 2-.beta.-cyanoethyl cyclohexanone, 2-b-cyanoethyl cyclohexanone, 2-oxocyclohexanepropiononitrile | |
O=C1CCCCC1CCC#N | |
151.21 | |
151.21 |
4594-78-9 | |
138°C to 142°C (10 mmHg) | |
MFCD00013797 | |
UN3276 | |
SPTVCXKSSRBTMN-UHFFFAOYNA-N | |
3-(2-oxocyclohexyl)propanenitrile | |
96223 | |
99% |
Chemical Identifiers
4594-78-9 | |
151.21 | |
SPTVCXKSSRBTMN-UHFFFAOYNA-N | |
96223 | |
O=C1CCCCC1CCC#N |
C9H13NO | |
MFCD00013797 | |
3-2-oxocyclohexyl propanenitrile, 2-2-cyanoethyl cyclohexanone, cyclohexanepropanenitrile, 2-oxo, 2-2-cyanethyl cyclohexanone, 2-oxo-1-cyclohexanepropionitrile, cyclohexanepropionitrile, 2-oxo, 2-oxocyclohexanepropionitrile, 2-.beta.-cyanoethyl cyclohexanone, 2-b-cyanoethyl cyclohexanone, 2-oxocyclohexanepropiononitrile | |
3-(2-oxocyclohexyl)propanenitrile |
Safety and Handling
GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 224-991-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only