missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Oxo-1-pyrrolidinyl)acetic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6626006

 View more versions of this product

Catalog No. AAH6626006



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(2-Oxo-1-pyrrolidinyl)acetic acid
C6H9NO3
5 g
2-oxo-1-pyrrolidineacetic acid, 2-2-oxopyrrolidin-1-yl acetic acid, 2-oxopyrrolidin-1-yl acetic acid, 2-oxo-pyrrolidin-1-yl-acetic acid, 2-oxo-1-pyrrolidinyl acetic acid, 1-pyrrolidineacetic acid, 2-oxo, 2-2-oxopyrrolidinyl acetic acid, piracetam impuirty d, 2-pyrrolidone acetic acid, acmc-209lb1
C1CC(=O)N(C1)CC(=O)O
143.142
143.14
Solid
53934-76-2
MFCD00962836
4342
JGPIWNNFLKDTSR-UHFFFAOYSA-N
2-(2-oxopyrrolidin-1-yl)acetic acid
3146687
97%

Chemical Identifiers

53934-76-2
143.142
JGPIWNNFLKDTSR-UHFFFAOYSA-N
3146687
C1CC(=O)N(C1)CC(=O)O
C6H9NO3
MFCD00962836
2-oxo-1-pyrrolidineacetic acid, 2-2-oxopyrrolidin-1-yl acetic acid, 2-oxopyrrolidin-1-yl acetic acid, 2-oxo-pyrrolidin-1-yl-acetic acid, 2-oxo-1-pyrrolidinyl acetic acid, 1-pyrrolidineacetic acid, 2-oxo, 2-2-oxopyrrolidinyl acetic acid, piracetam impuirty d, 2-pyrrolidone acetic acid, acmc-209lb1
2-(2-oxopyrrolidin-1-yl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only