missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Oxo-1-imidazolidinecarbonyl Chloride, TCI America™

Manufacturer:  TCI America O035225G

 View more versions of this product

Catalog No. O035225G



Specifications

Specifications

2-Oxo-1-imidazolidinecarbonyl Chloride
157°C
13214-53-4
MFCD02683529
NXJZQSRAFBHNLI-UHFFFAOYSA-N
2-oxoimidazolidine-1-carbonyl chloride
83241
Crystal-Powder at 20°C
White-Yellow
25g
C4H5ClN2O2
1-Chlorocarbonyl-2-imidazolidinone, 1-Chloroformyl-2-imidazolidinone
C1CN(C(=O)N1)C(=O)Cl
148.546
148.55
≥97.0% (T)

Chemical Identifiers

13214-53-4
148.546
NXJZQSRAFBHNLI-UHFFFAOYSA-N
83241
C1CN(C(=O)N1)C(=O)Cl
C4H5ClN2O2
MFCD02683529
1-Chlorocarbonyl-2-imidazolidinone, 1-Chloroformyl-2-imidazolidinone
2-oxoimidazolidine-1-carbonyl chloride
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents