missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Octyne 99.0+%, TCI America™

Manufacturer:  TCI America O012525ML

 View more versions of this product

Catalog No. O012525ML



Specifications

Specifications

2-Octyne
Colorless
2809-67-8
MFCD00027302
2-octyne
CCCCCC#CC
110.2
110.20
≥99.0% (GC)
137°C
25mL
C8H14
3295
QCQALVMFTWRCFI-UHFFFAOYSA-N
oct-2-yne
17769
Clear Liquid at 20°C

Chemical Identifiers

2809-67-8
110.2
QCQALVMFTWRCFI-UHFFFAOYSA-N
17769
CCCCCC#CC
C8H14
MFCD00027302
2-octyne
oct-2-yne
Safety and Handling

Safety and Handling

TSCA : No

Documents