Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Octyne, 98+%

Chemical Identifiers

Chemical Name or Material 2-Octyne
CAS 2809-67-8
Molecular Formula CH3(CH2)4C≡CCH3
Formula Weight 110.2
MDL Number MFCD00027302
UN Number UN3295
Beilstein 1697689
Synonym n-Pentyl methyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1019730

  SDS
alfa aesar™
A1019730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1019709

  SDS
alfa aesar™
A1019709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1019718

  SDS
alfa aesar™
A1019718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Octyne
CAS 2809-67-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Assay ≥98%
Molecular Formula CH3(CH2)4C≡CCH3
Formula Weight 110.2
MDL Number MFCD00027302
UN Number UN3295
Beilstein 1697689
Synonym n-Pentyl methyl acetylene
Boiling Point 137° to 138°C
Quantity 250g
Flash Point 28°C (82°F)
Odor Petroleum-like
Safety and Handling

Danger

GHS P Statement
P210-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.