missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Octene, cis + trans, 98%

Manufacturer:  Alfa Aesar™ L0386214

 View more versions of this product

Catalog No. AAL0386214



Specifications

Specifications

2-Octene
123°C to 125°C
Cis + Trans
C8H16
UN3295
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
CCCCCC=CC
112.216
112.22
14°C (57°F)
25g
111-67-1
MFCD00009532
1719495
ILPBINAXDRFYPL-HWKANZROSA-N
(E)-oct-2-ene
5364448
98%

Chemical Identifiers

111-67-1
112.216
ILPBINAXDRFYPL-HWKANZROSA-N
5364448
CCCCCC=CC
C8H16
MFCD00009532
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2
(E)-oct-2-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA

Documents