missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Octanoylthiophene, 99%

$37.84 - $445.78

Chemical Identifiers

CAS 30711-41-2
Molecular Formula C12H18OS
Formula Weight 210.34
MDL Number MFCD00053146
Beilstein 4220
Synonym n-Heptyl 2-thienyl ketone
Chemical Name or Material 2-Octanoylthiophene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2167814 SDS Alfa Aesar™
B2167814
25g Each for $135.53
AAB2167806 SDS Alfa Aesar™
B2167806
5g Each for $37.84
AAB2167822 SDS Alfa Aesar™
B2167822
100g Each for $445.78
Specifications Description & Specifications

Chemical Identifiers

30711-41-2
210.34
4220
2-Octanoylthiophene
C12H18OS
MFCD00053146
n-Heptyl 2-thienyl ketone

Specifications

30711-41-2
210.34
4220
120° to 122°C (1mmHg)
2-Octanoylthiophene
C12H18OS
MFCD00053146
n-Heptyl 2-thienyl ketone
25g
99%