Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(o-Tolyl)ethylamine, 98%

Chemical Identifiers

Chemical Name or Material 2-(o-Tolyl)ethylamine
CAS 55755-16-3
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD01310827
UN Number UN2735
Synonym 2-Methylphenethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2832003

  SDS
alfa aesar™
H2832003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2832006

  SDS
alfa aesar™
H2832006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.21
CAS 55755-16-3
Molecular Formula C9H13N
Chemical Name or Material 2-(o-Tolyl)ethylamine
Synonym 2-Methylphenethylamine
Quantity 1g
Assay 98%
MDL Number MFCD01310827
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
UN Number UN2735
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is employed as intermediate for pharmaceutical and organic synthesis

Solubility
Not miscible or difficult to mix with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.