missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(o-Tolyl)ethanol, TCI America™

$91.67 - $273.48

Chemical Identifiers

CAS 19819-98-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00044241
InChI Key RUGISKODRCWQNE-UHFFFAOYSA-N
Synonym 2-o-tolyl ethanol, 2-methylphenethyl alcohol, 2-2-methylphenyl ethanol, benzeneethanol, 2-methyl, o-methylphenethyl alcohol, phenethyl alcohol, o-methyl, 2-methylbenzeneethanol, toluene-ethanol, unii-vx8700jgjp, 2-2-methylphenyl ethan-1-ol
PubChem CID 88267
IUPAC Name 2-(2-methylphenyl)ethanol
SMILES CC1=CC=CC=C1CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13755G View Documents TCI America
M13755G
5g Each for $91.67
 
M137525G View Documents TCI America
M137525G
25g Each for $273.48
 
Specifications

Chemical Identifiers

19819-98-8
136.194
RUGISKODRCWQNE-UHFFFAOYSA-N
88267
CC1=CC=CC=C1CCO
C9H12O
MFCD00044241
2-o-tolyl ethanol, 2-methylphenethyl alcohol, 2-2-methylphenyl ethanol, benzeneethanol, 2-methyl, o-methylphenethyl alcohol, phenethyl alcohol, o-methyl, 2-methylbenzeneethanol, toluene-ethanol, unii-vx8700jgjp, 2-2-methylphenyl ethan-1-ol
2-(2-methylphenyl)ethanol

Specifications

243°C
5g
C9H12O
2-o-tolyl ethanol, 2-methylphenethyl alcohol, 2-2-methylphenyl ethanol, benzeneethanol, 2-methyl, o-methylphenethyl alcohol, phenethyl alcohol, o-methyl, 2-methylbenzeneethanol, toluene-ethanol, unii-vx8700jgjp, 2-2-methylphenyl ethan-1-ol
CC1=CC=CC=C1CCO
136.194
136.19
≥98.0% (GC)
Yellow
19819-98-8
MFCD00044241
RUGISKODRCWQNE-UHFFFAOYSA-N
2-(2-methylphenyl)ethanol
88267
Clear Liquid at 20°C
2-(o-Tolyl)ethanol
Safety and Handling

Safety and Handling

TSCA : Yes