Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-O-Methylinosine, 99%

Chemical Identifiers

Chemical Name or Material 2'-O-Methylinosine
CAS 3881-21-8
Molecular Formula C11H14N4O5
Formula Weight 282.25
MDL Number MFCD01631008
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64748MD

  SDS
alfa aesar™
J64748MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 282.25
Chemical Name or Material 2'-O-Methylinosine
CAS 3881-21-8
Physical Form Powder
Assay 99%
Molecular Formula C11H14N4O5
MDL Number MFCD01631008
Color White to Off-white
Quantity 250mg
Recommended Storage 2° to 8°C

Applications
2′-O-Methylinosine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

2° to 8°C