Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nonyne, 98%

Chemical Identifiers

Chemical Name or Material 2-Nonyne
CAS 19447-29-1
Molecular Formula C9H16
Linear Formula CH3C≡C(CH2)5CH3
Formula Weight 124.23
MDL Number MFCD00039986
UN Number UN3295
Synonym Methyl n-hexyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2362006

  SDS
alfa aesar™
B2362006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2362014

  SDS
alfa aesar™
B2362014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.23
Health Hazard 3 GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
Chemical Name or Material 2-Nonyne
Health Hazard 2 GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Molecular Formula C9H16
Boiling Point 158° to 159°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Assay 98%
UN Number UN3295
CAS 19447-29-1
Health Hazard 1 Danger
Quantity 5g
Linear Formula CH3C≡C(CH2)5CH3
Synonym Methyl n-hexyl acetylene
MDL Number MFCD00039986
Safety and Handling

Danger

GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.