Please login to your online account to display your discounted pricing

2-Nonanol 99%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  129150100

 View more versions of this product

Catalog No. AC129150100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 144.26
Quantity 10mL
Color Clear yellow
EINECS Number 211-065-1
TSCA TSCA
Density 0.8200g/mL
CAS Max % 100.0
Chemical Name or Material 2-Nonanol
Packaging Glass bottle
Name Note 99%
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C9H20O
Linear Formula CH3(CH2)6CH(OH)CH3
MDL Number MFCD00004593
CAS 628-99-9
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Nonanol
Name Note 99%
CAS 628-99-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H20O
Linear Formula CH3(CH2)6CH(OH)CH3
Formula Weight 144.26
MDL Number MFCD00004593
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents