Please login to your online account to display your discounted pricing

2-Nonanol 99%, ACROS Organics™

$27.25 - $27.25

Chemical Identifiers

Chemical Name or Material 2-Nonanol
Name Note 99%
CAS 628-99-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H20O
Linear Formula CH3(CH2)6CH(OH)CH3
Formula Weight 144.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC129150100

  SDS
thermo fisher scientific
129150100
Glass bottle 10mL Each for $27.25
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 144.26
Quantity 10mL
Color Clear yellow
EINECS Number 211-065-1
TSCA TSCA
Density 0.8200g/mL
CAS Max % 100.0
Chemical Name or Material 2-Nonanol
Packaging Glass bottle
Name Note 99%
Refractive Index 1.4290 to 1.4310 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C9H20O
Linear Formula CH3(CH2)6CH(OH)CH3
MDL Number MFCD00004593
CAS 628-99-9
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Infrared Spectrum Authentic
Safety and Handling

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.