Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nonanol, 99%

$57.73 - $146.85

Chemical Identifiers

Chemical Name or Material 2-Nonanol
CAS 628-99-9
Molecular Formula C9H20O
Formula Weight 144.26
MDL Number MFCD00004593
Beilstein 1719466
Synonym n-Heptyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0631014

  SDS
alfa aesar™
L0631014
25g Each for $57.73

AAL0631022

  SDS
alfa aesar™
L0631022
100g Each for $146.85
Description & Specifications

Specifications

Formula Weight 144.26
Chemical Name or Material 2-Nonanol
MDL Number MFCD00004593
CAS 628-99-9
Synonym n-Heptyl methyl carbinol
Health Hazard 1 Warning
TSCA TSCA
Assay 99%
Molecular Formula C9H20O
Beilstein 1719466
Boiling Point 195° to 197°C
Melting Point −35°C
Quantity 25g
Flash Point 82°C (179°F)
Safety and Handling

Warning