Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrothiophene-4-carboxylic acid, 98%

$191.36 - $191.36

Chemical Identifiers

Chemical Name or Material 2-Nitrothiophene-4-carboxylic acid
CAS 40357-96-8
Molecular Formula C5H3NO4S
Formula Weight 173.15
MDL Number MFCD00052582
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1070503

  SDS
alfa aesar™
A1070503
1g Each for $191.36
Description & Specifications

Specifications

Formula Weight 173.15
CAS 40357-96-8
MDL Number MFCD00052582
Assay 98%
Melting Point 145° to 148°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C5H3NO4S
Chemical Name or Material 2-Nitrothiophene-4-carboxylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.