Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitrothiophene-4-carboxaldehyde, 97%

$73.26 - $224.73

Chemical Identifiers

Chemical Name or Material 2-Nitrothiophene-4-carboxaldehyde
CAS 75428-45-4
Molecular Formula C5H3NO3S
Formula Weight 157.15
MDL Number MFCD00052581
Beilstein 1342966
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1079303

  SDS
alfa aesar™
L1079303
1g Each for $73.26

AAL1079306

  SDS
alfa aesar™
L1079306
5g Each for $224.73
Description & Specifications

Specifications

Formula Weight 157.15
Chemical Name or Material 2-Nitrothiophene-4-carboxaldehyde
Assay 97%
Molecular Formula C5H3NO3S
CAS 75428-45-4
Beilstein 1342966
MDL Number MFCD00052581
Melting Point 79° to 81°C
Quantity 1g